Ethyl-8-iodo-2,6-bis(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2,6-bis(trifluoromethyl)-8-iodoimidazo[1,2-a]pyridine-3-carboxylate
046987-500MG
101877-914EA
921.54
USD
101877-914
Ethyl-8-iodo-2,6-bis(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-iodo-2,6-bis(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₂H₇F₆IN₂O₂ Melting Pt: 167…168 °C |
MDL Number:
MFCD12498697 CAS Number: 1171918-86-7 |
Specification Test Results
Melting point | 167-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...