6-Bromo-1H-indole-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1H-indole-2-carboxaldehyde

046985-500MG
101877-908EA 389.23 USD
101877-908
6-Bromo-1H-indole-2-carbaldehyde ≥95%
6-Bromo-1H-indole-2-carbaldehyde

Hazard: Irritant

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 181…183 °C
MDL Number: MFCD06738308
CAS Number: 105191-12-6

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR