Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

046981-500MG 046981-1G
101877-894EA 1296.92 USD
101877-894 101877-896
Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD12025931
CAS Number: 1140512-58-8

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR