Methyl-3-amino-6-nitro-1-benzothiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
046963-1G
046963-5G
046963-500MG
101877-822EA
786.15
USD
101877-822
101877-824
101877-820
Methyl-3-amino-6-nitro-1-benzothiophene-2-carboxylate ≥95%
Methyl-3-amino-6-nitro-1-benzothiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈N₂O₄S Melting Pt: 231…232 °C |
MDL Number:
MFCD02647200 CAS Number: 35212-90-9 |
Specification Test Results
Melting point | 231-232°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...