6-Chloro-5-methylnicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-methylpyridine-3-carbaldehyde

046949-500MG 046949-1G
101877-762EA 53.85 USD
101877-762 101877-764
6-Chloro-5-methylnicotinaldehyde ≥95%
6-Chloro-5-methylnicotinaldehyde

Hazard: Irritant

Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD11520655
CAS Number: 176433-43-5

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR