2-Chloro-6-iodo-3-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-iodopyridin-3-ol

046945-1G 046945-5G
101877-744EA 46.15 USD
101877-744 101877-746
2-Chloro-6-iodo-3-pyridinol ≥95%
2-Chloro-6-iodo-3-pyridinol

Hazard: Irritant

Formula: C₅H₃ClINO
MW: 255.44 g/mol
Melting Pt: 142…143 °C
MDL Number: MFCD10566651
CAS Number: 185220-68-2

Order Now

Specification Test Results

Melting point 142-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR