Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxy-6-(trifluoromethyl)-1H-indole

046942-500MG 046942-1G
101877-730EA 556.92 USD
101877-730 101877-732
Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether ≥95%
Methyl-6-(trifluoromethyl)-1H-indol-5-yl ether

Hazard: Irritant

Formula: C₁₀H₈F₃NO
MW: 215.18 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD11848569
CAS Number: 178896-78-1

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR