4-Amino-1-isoindolinone ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2,3-dihydro-1H-isoindol-1-one
046931-1G
046931-500MG
101877-682EA
218.46
USD
101877-682
101877-684
4-Amino-1-isoindolinone ≥95%
4-Amino-1-isoindolinone
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.17 g/mol Melting Pt: 200…202 °C |
MDL Number:
MFCD08234883 CAS Number: 366452-98-4 |
Specification Test Results
Melting point | 200-202°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...