tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
N'-Benzylsulphamide, N-BOC protected
046917-1G
046917-5G
046917-500MG
101877-624EA
115.38
USD
101877-624
101877-626
101877-628
tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate ≥95%
tert-Butyl-3-benzyl-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₄S MW: 286.35 g/mol Melting Pt: 106…108 °C |
MDL Number:
MFCD09864818 CAS Number: 147000-78-0 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...