Ethyl-1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

046915-1G 046915-5G 046915-500MG
101877-618EA 181.54 USD
101877-618 101877-620 101877-622
Ethyl-1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O₃
MW: 285.73 g/mol
Melting Pt: 174…175 °C
MDL Number: MFCD09864820
CAS Number: 952183-76-5

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR