4-Fluoro-N-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046906-500MG
046906-5G
046906-1G
101877-568EA
181.54
USD
101877-568
101877-570
101877-572
4-Fluoro-N-methylbenzenesulfonamide ≥95%
4-Fluoro-N-methylbenzenesulphonamide
Hazard: Irritant
Formula:
C₇H₈FNO₂S MW: 189.2073 g/mol Melting Pt: 56…58 °C |
MDL Number:
MFCD01211989 CAS Number: 433-14-7 |
Specification Test Results
Melting point | 56-58°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...