4-Methoxy-N-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046905-1G
046905-500MG
046905-5G
101877-562EA
150.77
USD
101877-562
101877-564
101877-566
4-Methoxy-N-methylbenzenesulfonamide ≥95%
4-Methoxy-N-methylbenzenesulphonamide
Hazard: Irritant
Formula:
C₈H₁₁NO₃S Melting Pt: 98…100 °C |
MDL Number:
MFCD01046356 CAS Number: 7010-86-8 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...