2-Chloro-N-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046897-1G
046897-500MG
046897-5G
101877-524EA
776.92
USD
101877-524
101877-520
101877-522
2-Chloro-N-methylbenzenesulfonamide ≥95%
2-Chloro-N-methylbenzenesulphonamide
Hazard: Irritant
Formula:
C₇H₈ClNO₂S Melting Pt: 94…96 °C |
MDL Number:
MFCD00859227 CAS Number: 17260-67-2 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...