Methyl-4-[4-(tert-butyl)phenyl]-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate ≥95%
Supplier: Matrix Scientific
046894-500MG
046894-5G
046894-1G
101877-502EA
181.54
USD
101877-502
101877-504
101877-506
Methyl-4-[4-(tert-butyl)phenyl]-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate ≥95%
Methyl-4-[4-(tert-butyl)phenyl]-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate
Hazard: Irritant
Formula:
C₁₉H₂₄N₂O₅ MW: 360.41 g/mol Melting Pt: 231…232 °C |
MDL Number:
MFCD09817439 CAS Number: 952183-65-2 |
Specification Test Results
Melting point | 231-232°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...