4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid

046887-500MG 046887-1G 046887-5G
101877-462EA 181.54 USD
101877-462 101877-464 101877-466
4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid ≥95%
4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD00052170
CAS Number: 56671-28-4

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR