Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

046876-500MG 046876-1G 046876-5G
101877-408EA 375.38 USD
101877-408 101877-410 101877-412
Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate ≥95%
Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₉BrN₂O₂
MW: 293.13 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD08444032
CAS Number: 924869-15-8

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR