4-[4-(1-Pyrrolidinyl)phenyl]-1,3-thiazol-2-ylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-4-(4-pyrrolidin-1-ylphenyl)-1,3-thiazole
046867-1G
046867-500MG
101877-360EA
227.69
USD
101877-360
101877-362
4-[4-(1-Pyrrolidinyl)phenyl]-1,3-thiazol-2-ylamine ≥95%
4-[4-(1-Pyrrolidinyl)phenyl]-1,3-thiazol-2-ylamine
Hazard: Irritant
Formula:
C₁₃H₁₅N₃S MW: 245.35 g/mol Melting Pt: 228…230 °C |
MDL Number:
MFCD08689684 CAS Number: 784136-89-6 |
Specification Test Results
Melting point | 228-230°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...