Ethyl-4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-[4-(methoxycarbonyl)phenyl]piperazine-1-carboxylate
046863-1G
046863-500MG
046863-5G
101877-338EA
181.54
USD
101877-338
101877-340
101877-342
Ethyl-4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-[4-(methoxycarbonyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₄ MW: 292.34 g/mol Melting Pt: 109…111 °C |
MDL Number:
MFCD08689682 CAS Number: 924869-10-3 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...