1-(2-Nitrophenyl)-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
046860-500MG
046860-1G
046860-5G
101877-326EA
806.15
USD
101877-326
101877-322
101877-324
1-(2-Nitrophenyl)-1H-pyrrole-2-carbaldehyde ≥95%
1-(2-Nitrophenyl)-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₁H₈N₂O₃ MW: 216.2 g/mol Melting Pt: 124…126 °C |
MDL Number:
MFCD00482264 CAS Number: 33265-61-1 |
Specification Test Results
Melting point | 124-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...