5-Nitro-1,2-dihydro-3H-indazol-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Dihydro-5-nitro-3H-indazol-3-one

046853-500MG 046853-5G 046853-1G
101877-282EA 195.97 USD
101877-282 101877-284 101877-286
5-Nitro-1,2-dihydro-3H-indazol-3-one ≥95%
5-Nitro-1,2-dihydro-3H-indazol-3-one

Hazard: Irritant

Formula: C₇H₅N₃O₃
MW: 179.14 g/mol
Melting Pt: 280…282 °C
MDL Number: MFCD00012418
CAS Number: 61346-19-8

Order Now

Specification Test Results

Melting point 280-282°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR