2-(6-Chloro-3-pyridinyl)-1,3-benzoxazole ≥95%
Supplier: Matrix Scientific
046846-500MG
046846-5G
046846-1G
101877-242EA
556.92
USD
101877-242
101877-244
101877-246
2-(6-Chloro-3-pyridinyl)-1,3-benzoxazole ≥95%
2-(6-Chloro-3-pyridinyl)-1,3-benzoxazole
Hazard: Irritant
Formula:
C₁₂H₇ClN₂O MW: 230.66 g/mol Melting Pt: 177…179 °C |
MDL Number:
MFCD08689673 CAS Number: 54628-03-4 |
Specification Test Results
Melting point | 177-179°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...