2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole ≥95%
Supplier: Matrix Scientific
046845-500MG
046845-5G
046845-1G
101877-238EA
698.46
USD
101877-238
101877-240
101877-236
2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole ≥95%
2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole
Hazard: Irritant
Formula:
C₁₂H₇ClN₂O MW: 230.66 g/mol Melting Pt: 94…95 °C |
MDL Number:
MFCD08689672 CAS Number: 134673-71-5 |
Specification Test Results
Melting point | 94-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...