5-(1,3-Benzoxazol-2-yl)-2-pyridinol ≥95%
Supplier: Matrix Scientific
Synonyms:
5-(1,3-Benzoxazol-2-yl)-2-hydroxypyridine
046844-1G
046844-5G
046844-500MG
101877-230EA
181.54
USD
101877-230
101877-232
101877-234
5-(1,3-Benzoxazol-2-yl)-2-pyridinol ≥95%
5-(1,3-Benzoxazol-2-yl)-2-pyridinol
Hazard: Irritant
Formula:
C₁₂H₈N₂O₂ MW: 212.21 g/mol Melting Pt: 280…282 °C |
MDL Number:
MFCD08444021 CAS Number: 54627-93-9 |
Specification Test Results
Melting point | 280-282°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...