(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol ≥95%

Supplier: Matrix Scientific

046840-500MG 046840-1G
101877-210EA 556.92 USD
101877-210 101877-212
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol ≥95%
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol

Hazard: Irritant

Formula: C₅H₆Br₂N₂O
Melting Pt: 114…116 °C
MDL Number: MFCD12025921
CAS Number: 881997-90-6

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR