[4-(1,3-Oxazol-5-yl)phenyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: [4-(5-Oxazolyl)phenyl]methanol5-[4-(Hydroxymethyl)phenyl]-1,3-oxazole

046835-500MG
101877-190EA 661.54 USD
101877-190
[4-(1,3-Oxazol-5-yl)phenyl]methanol ≥95%
[4-(1,3-Oxazol-5-yl)phenyl]methanol

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD02682042
CAS Number: 179057-18-2

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR