4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
046834-500MG
101877-188EA
1296.92
USD
101877-188
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₃ Melting Pt: 225…227 °C |
MDL Number:
MFCD12025919 CAS Number: 532391-92-7 |
Specification Test Results
Melting point | 225-227°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...