6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one ≥95%
Supplier: Matrix Scientific
046828-1G
046828-500MG
101877-170EA
472.31
USD
101877-170
101877-172
6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one ≥95%
6-Bromo-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one
Hazard: Irritant
Formula:
C₇H₅BrN₂O₂ MW: 229.04 g/mol Melting Pt: 237…239 °C |
MDL Number:
MFCD12025916 CAS Number: 335032-38-7 |
Specification Test Results
Melting point | 237-239°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...