(2,5-Difluoro-4-nitrophenyl)methanol ≥97%

Supplier: Matrix Scientific

046817-500MG 046817-1G
101877-128EA 375.38 USD
101877-128 101877-130
(2,5-Difluoro-4-nitrophenyl)methanol ≥97%
(2,5-Difluoro-4-nitrophenyl)methanol

Hazard: Irritant

Formula: C₇H₅F₂NO₃
Melting Pt: 80…81 °C
MDL Number: MFCD09909606
CAS Number: 1160474-68-9

Order Now

Specification Test Results

Melting point 80-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR