2-(Acetylamino)-5-bromo-3-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

046813-500MG
101877-120EA 747.69 USD
101877-120
2-(Acetylamino)-5-bromo-3-thiophenecarboxylic acid ≥95%
2-(Acetylamino)-5-bromo-3-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₇H₆BrNO₃S
Melting Pt: 220…222 °C
MDL Number: MFCD12025910
CAS Number: 1160474-66-7

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR