3-Amino-3,4-dihydroquinolin-2(1H)-one hydrochloride ≥95%
Supplier: Matrix Scientific
046812-1G
046812-5G
046812-500MG
101877-118EA
3000
USD
101877-118
101877-114
101877-116
3-Amino-3,4-dihydroquinolin-2(1H)-one hydrochloride ≥95%
3-Amino-3,4-dihydroquinolin-2(1H)-one hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O Melting Pt: 314…317 °C (dec.) |
MDL Number:
MFCD09999258 CAS Number: 40615-17-6 |
Specification Test Results
Melting point | 314-317°C(d |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...