N-(3-Chloro-5-nitro-2-pyridinyl)-N-ethylamine ≥95%
Supplier: Matrix Scientific
046806-500MG
046806-1G
101877-098EA
375.38
USD
101877-098
101877-100
N-(3-Chloro-5-nitro-2-pyridinyl)-N-ethylamine ≥95%
N-(3-Chloro-5-nitro-2-pyridinyl)-N-ethylamine
Hazard: Irritant
Formula:
C₇H₈ClN₃O₂ Melting Pt: 98…100 °C |
MDL Number:
MFCD12025905 CAS Number: 1160474-57-6 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...