1-[4-Nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
046793-5G
046793-500MG
046793-1G
101877-032EA
278.46
USD
101877-032
101877-034
101877-036
1-[4-Nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylic acid ≥95%
1-[4-Nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₃F₃N₂O₄ Melting Pt: 147…149 °C |
MDL Number:
MFCD11039558 CAS Number: 1171918-77-6 |
Specification Test Results
Melting point | 147-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...