2-[(Benzylamino)carbonyl]cyclopropanecarboxylic acid ≥95%

Supplier: Matrix Scientific

046792-500MG
101877-030EA 747.69 USD
101877-030
2-[(Benzylamino)carbonyl]cyclopropanecarboxylic acid ≥95%
2-[(Benzylamino)carbonyl]cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
Melting Pt: 93…94 °C
MDL Number: MFCD12025898
CAS Number: 1160474-47-4

Order Now

Specification Test Results

Melting point 93-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR