Methyl-3-amino-2,2-dimethylpropanoate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-amino-2,2-dimethylpropanoate hydrochloride
046784-500MG
101876-996EA
747.69
USD
101876-996
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride ≥95%
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride
Hazard: Irritant
Formula:
C₆H₁₄ClNO₂ MW: 167.64 g/mol Melting Pt: 167…168 °C |
MDL Number:
MFCD10758092 CAS Number: 177269-37-3 |
Specification Test Results
Melting point | 167-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...