Ethyl-6-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
046777-1G
046777-500MG
101876-968EA
556.92
USD
101876-968
101876-970
Ethyl-6-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-chloro-8-iodoimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈ClIN₂O₂ Melting Pt: 129…130 °C |
MDL Number:
MFCD10758086 CAS Number: 1033463-34-1 |
Specification Test Results
Melting point | 129-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...