1-Cyanocyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

046772-500MG 046772-1G
101876-946EA 403.08 USD
101876-946 101876-948
1-Cyanocyclobutanecarboxylic acid ≥95%
1-Cyanocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₆H₇NO₂
Melting Pt: 68…69 °C
MDL Number: MFCD11100208
CAS Number: 30491-91-9

Order Now

Specification Test Results

Melting point 68-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR