[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol ≥90%

Supplier: Matrix Scientific

Synonyms: (3-Chloro-5-(trifluoromethyl)pyridin-2-yl)methanol3-Chloro-2-(hydroxymethyl)-5-(trifluoromethyl)pyridine

046768-1G
101876-932EA 484.62 USD
101876-932
[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol ≥90%
[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol

Hazard: Irritant

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Melting Pt: 36…38 °C
MDL Number: MFCD11100207
CAS Number: 1033463-31-8

Order Now

Specification Test Results

Melting point 36-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR