2-Amino-5-iodo-3-pyridinylamine ≥95%

Supplier: Matrix Scientific

046764-500MG
101876-918EA 435.38 USD
101876-918
2-Amino-5-iodo-3-pyridinylamine ≥95%
2-Amino-5-iodo-3-pyridinylamine

Hazard: Irritant

Formula: C₅H₆IN₃
Melting Pt: 109…111 °C
MDL Number: MFCD10697693
CAS Number: 426463-01-6

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR