Ethyl-8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
046759-500MG
046759-1G
101876-898EA
615.38
USD
101876-898
101876-896
Ethyl-8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 8-amino-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O₂ Melting Pt: 150…152 °C |
MDL Number:
MFCD10758082 CAS Number: 185133-90-8 |
Specification Test Results
Melting point | 150-152°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...