5-Amino-1-(6-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
046756-500MG
046756-1G
046756-5G
101876-882EA
269.23
USD
101876-882
101876-886
101876-884
5-Amino-1-(6-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(6-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile
Hazard: Irritant
Formula:
C₉H₆ClN₅ MW: 219.63 g/mol Melting Pt: 220…221 °C |
MDL Number:
MFCD01936342 CAS Number: 400084-55-1 |
Specification Test Results
Melting point | 220-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...