1-[2-(3-Pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

046747-1G 046747-500MG 046747-5G
101876-846EA 698.46 USD
101876-846 101876-844 101876-848
1-[2-(3-Pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(3-Pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₀H₉N₃OS
Melting Pt: 230…232 °C
MDL Number: MFCD09607950
CAS Number: 952183-61-8

Order Now

Specification Test Results

Melting point 230-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR