1-[4-Methyl-2-(3-pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
Supplier: Matrix Scientific
046746-5G
046746-1G
046746-500MG
101876-838EA
375.38
USD
101876-838
101876-842
101876-840
1-[4-Methyl-2-(3-pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[4-Methyl-2-(3-pyridinylamino)-1,3-thiazol-5-yl]-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₁₁N₃OS Melting Pt: 208…210 °C |
MDL Number:
MFCD09607949 CAS Number: 952183-60-7 |
Specification Test Results
Melting point | 208-210°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...