1-[2-(2,5-Dimethoxyanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Acetyl-2-[(2,5-dimethoxyphenyl)amino]-4-methyl-1,3-thiazole
046744-500MG
046744-1G
046744-5G
101876-828EA
375.38
USD
101876-828
101876-832
101876-830
1-[2-(2,5-Dimethoxyanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(2,5-Dimethoxyanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone
Hazard: Irritant
Formula:
C₁₄H₁₆N₂O₃S Melting Pt: 140…142 °C |
MDL Number:
MFCD02110102 CAS Number: 398472-44-1 |
Specification Test Results
Melting point | 140-142°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...