Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-[2-(2,5-Dimethoxyanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific


046743-500MG 046743-5G 046743-1G
101876-822EA 473.38 USD InStock
101876-822 101876-826 101876-824
1-[2-(2,5-Dimethoxyanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(2,5-Dimethoxyanilino)-1,3-thiazol-5-yl]-1-ethanone
Purity: 95%
CAS: 952183-59-4
MDL Number: MFCD09607948
Molecular Formula: C13H14N2O3S
Molecular Weight: 278,33
Hazard: Irritant


Formula: C₁₃H₁₄N₂O₃S
Melting Pt: 150…152 °C
MDL Number: MFCD09607948
CAS Number: 952183-59-4
EINECS: 000-000-0
Order Now
Specification Test Results
Melting point 150-152°C


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty