4-Cyano-3-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-formylbenzonitrile4-Cyano-3-fluorobenzaldehyde

046736-1G
101876-792EA 71.3 USD
101876-792
4-Cyano-3-fluorobenzaldehyde ≥95%
4-Cyano-3-fluorobenzaldehyde

Hazard: Irritant

Formula: C₈H₄FNO
MW: 149.1200 g/mol
Melting Pt: 89…91 °C
MDL Number: MFCD06738989
CAS Number: 101048-76-4

Order Now

Specification Test Results

Melting point 89-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR