Ethyl-5-cyano-2-[4-(2-hydroxyethyl)piperazino]-6-methylnicotinate ≥95%

Supplier: Matrix Scientific

046722-1G 046722-500MG 046722-5G
101876-736EA 556.92 USD
101876-736 101876-738 101876-740
Ethyl-5-cyano-2-[4-(2-hydroxyethyl)piperazino]-6-methylnicotinate ≥95%
Ethyl-5-cyano-2-[4-(2-hydroxyethyl)piperazino]-6-methylnicotinate

Hazard: Irritant

Formula: C₁₆H₂₂N₄O₃
MW: 318.38 g/mol
Melting Pt: 95…97 °C
MDL Number: MFCD08444015
CAS Number: 924869-05-6

Order Now

Specification Test Results

Melting point 95-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR