Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate ≥95%

Supplier: Matrix Scientific

046709-5G 046709-500MG 046709-1G
101876-668EA 556.92 USD
101876-668 101876-670 101876-672
Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate ≥95%
Methyl 2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₁BrN₂O₂
Melting Pt: 134…136 °C
MDL Number: MFCD08444011
CAS Number: 133427-41-5

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR