1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

046708-500MG 046708-5G 046708-1G
101876-662EA 556.92 USD
101876-662 101876-666 101876-664
1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid ≥95%
1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₃
MW: 261.32 g/mol
Melting Pt: 278…281 °C
MDL Number: MFCD03645498
CAS Number: 887440-34-8

Order Now

Specification Test Results

Melting point 278-281°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR