3-Iodo-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Iodopyrazole

046651-1G
101876-416EA 107.69 USD
101876-416
3-Iodo-1H-pyrazole ≥95%
3-Iodo-1H-pyrazole

Hazard: Irritant

Formula: C₃H₃IN₂
MW: 193.98 g/mol
Melting Pt: 74…75 °C
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now

Specification Test Results

Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR