5-Bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046649-500MG 046649-1G
101876-406EA 921.54 USD
101876-406 101876-408
5-Bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylic acid ≥95%
5-Bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₄BrNO₅
Melting Pt: >205 °C (dec.)
MDL Number: MFCD12025882
CAS Number: 1160474-43-0

Order Now

Specification Test Results

Melting point 205°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR